Anti-LGI1 Mouse Monoclonal Antibody (ALP) [Clone: S283-7]

Supplier: Rockland Immunochemicals
200-305-W73
75929-588EA 539 USD
75929-588
Anti-LGI1 Mouse Monoclonal Antibody (ALP) [Clone: S283-7]
Antibodies
The leucine-rich, glioma inactivated gene 1 (LGI1) was first identified as a candidate tumor suppressor gene for glioma and may play a role in other cancers. LGI1 is a member of a family of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 serves as a ligand to ADAM22, a metalloprotease localized at the synapse. Mutations in LGI1 account for nearly half of autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Two isoforms of LGI1 are known to exist; this LGI1 antibody will recognize only the longer form. Anti-LGI1 is ideal for research in Neuroscience and Cancer.

Anti-LGI1 Alkaline Phosphatase Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~60kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: LGI1
Clonality: Monoclonal
Clone: S283-7
Conjugation: ALP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR