VHB™ Adhesive Transfer Tapes, TapeCase
Supplier: 3M
F9460PC-0.1875 7000048539 F9460PC-1/4 F-9460PC-1
76242-180EA 180.93 USD
76242-180 76243-642 76248-112 76243-142
VHB™ Adhesive Transfer Tapes, TapeCase
Tapes
Adhesive transfer tapes are rolls of pressure sensitive adhesive pre-applied to a special release liner. The tape is simply pressed, adhesive side down, to a surface and the liner is peeled off.
  • 2.0 mil 100 MP adhesive
  • 4.2 mil 58 lbs. polycoated kraft paper liner
  • Suitable for industrial joining and metal fabrication
Used for bonding: decorative metal trim, flexible circuits to aluminum rigidizers or heat sinks.
Certifications: UL listing 746C