SnapLok™ Adapter Assemblies, Nordson EFD
Supplier: Nordson EFD
7016113 7016127 7016059 7016136 7016135 7016122 7016129 7005011 7018522
76237-762EA 107.15 USD
76237-762 76243-746 76241-998 76237-760 76240-422 76242-000 76241-992 76250-986 76245-026
SnapLok™ Adapter Assemblies, Nordson EFD
Syringe Barrels, Plungers and Pistons
One-piece molded SnapLok™ adapter provides safe attachment of syringe barrel.
Delivery information: Includes 5/32" OØ flexible polyurethane tubing, Buna N O-ring, quick-connect and safety clip.