Dichloromethane ≥99.0%, GenAR® NF, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Methane dichloride
, Methylene chloride
Management of Change (MOC) category = R
MACR4874-QM
MACR4874-26
MACR4874-26
MK4874-26EA
18557.26
USD
MK4874-26
JT4874-QM
CAMK4874-26
Dichloromethane ≥99.0%, GenAR® NF, Macron Fine Chemicals™
Dichloromethane
Formula:
CH₂Cl₂ MW: 84.93 g/mol Density: 1.32 g/cm³ |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 UN: 1593 ADR: 6.1,III |
Specification Test Results
Meets E.P. Chemical Specifications | |
GMP Manufactured Product | |
Meets N.F. Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Assay | ≥99.0 % |
NF - Identification | Passes Test |
NF - Free Chlorine (Cl₂) | Passes Test |
NF - Hydrogen Chloride (HCl) | ≤0.001 % |
NF - Methanol | ≤600 ppm |
NF - Nonvolatile Residue | ≤0.002 % |
NF - Specific Gravity at 25°C | 1.318 - 1.322 |
NF - Water | ≤0.02 % |
1,1-dichloroethene | |
EP - Appearance | Passes Test |
EP - Refractive Index | 1.423 - 1.425 Degree |
EP - Identification B | Passes Test |
EP - Identification C | Passes Test |
EP - Free Chlorine | Passes Test |
EP - Acidity | Passes Test |
EP - Residue on Evaporation | ≤20 ppm |
EP - Relative Density @ 20° C | 1.320 - 1.332 |
EP - Water (KF) | ≤0.02 % |
EP - Ethanol, v/v | ≤2.0 % |
EP - 2-methylbut-2-ene, v/v | ≤300 ppm |
EP - Carbon tetrachloride, v/v | ≤10 ppm |
EP - Chloroform, v/v | ≤50 % |
EP - Total other impurities v/v | ≤0.1 % |
Residual Solvents - The Class 1 solvent 1,1-dichloroethene is likely to | |
be present. The concentration of 1,1-dichloroethene is quantified and | |
reported for each batch. Methanol and amylene (2-methyl-butylene) are | |
intentionally added as stabilizers. The concentration of methanol is | |
reported in the batch results. The concentration of amylene is less | |
than 40 ppm based on in-process controls. The Class 2 solvents | |
cyclohexane (less than 20 ppm) and 1,2-dichloroethene (less than 50 ppm) may be present due to their presence in the process raw material. They | |
are not selectively removed during processing. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...