FM-203 Dual Port Soldering System, American Hakko Products
Supplier: American Hakko Products
FM203-01
76242-126EA 2011.36 USD
76242-126
FM-203 Dual Port Soldering System, American Hakko Products
Soldering Supplies Soldering Stations
Dual port soldering system with one soldering handpiece features to deliver heat to the soldering area fast. The dual soldering ports allow for a variety of soldering tools including SMD hot tweezers and desoldering tools.
  • Temperature accuracy ±15 °C
  • LED in handpiece tells the operator the iron is hot
  • Auto shutoff
  • Sleep mode functionality
  • No calibration necessary
Temperature range 200 - 450 °C (400 - 840 °F).
Ordering information: Tips has to be ordered separately. Supplied with soldering station, soldering iron, power cord, control card, green sleeve assembly (anto-bacterial), heat resistant pad, iron holder, connecting cable, tip tray and instruction manual.