Heating Gun HL 2020 E, Steinel America
Supplier: Steinel
HL 2020 E
76253-098EA 878.67 USD
76253-098
Heating Gun HL 2020 E, Steinel America
Heaters Hot Air Dryers
Heating gun provides convenient handling with its LCD display.
  • Ceramic heating element
  • One year limited Warranty
The residual heat indicator warns the user that the outlet nozzle is still hot. The precision adjustable heat and powerful blower make the HL 2020 E the ideal choice for virtually any application – heat shrinking tubes and sleeves onto cables, welding and many other challenging jobs.