IntelliTemp™ Heat Gun, Steinel America
Supplier: Steinel
34850
76246-032EA 658.24 USD
76246-032
IntelliTemp™ Heat Gun, Steinel America
Heaters Hot Air Dryers
Microprocessor controlled heat gun with LCD display enabling temperature selection in 10°F increments. Exclusive
DuraTherm™ heating element ensures long life and even heat.
  • One year limited warranty
  • Available with or without a heavy duty carrying case
Air flow: 3.6 / 10.6 / 17.6 CFM
Accessories information: Accepts a full range of nozzles and accessories.