Reduction Nozzle, Steinel America
Supplier: STEINEL AMERICA INC. TE
7062 7071
76242-504EA 33.34 USD
76242-504 76246-060
Reduction Nozzle, Steinel America
Heaters Hot Air Dryers
Provides a concentrated stream of hot air for precision.
  • Welding or soldering
  • May be used as a base for specialized attachments
  • Use on electronic heat guns only