KWIK-STIK Aspergillus terreus ATCC® 1012™, Microbiologics
Supplier: MICROBIOLOGICS, INC.
01264K
01264P
76299-410PK
266.94
USD
76299-410
76299-414
KWIK-STIK Aspergillus terreus ATCC® 1012™, Microbiologics
Controls and Calibration Verification Materials
Everything you need to grow reference cultures for QC testing is included in this one handy device.
- Over 900 strains available
- All-in-one design reduces the risk of contamination
- Strains are less than or equal to four passages from reference culture
- Ready-to-use format saves time and money
- Refrigerated storage is easy and economical
- Online Certificate of Analysis provides detailed strain information
- Traceability to reference cultures ensures authenticity
KWIK-STIK’s simple, all-inclusive design makes life easier for lab techs while reducing chances for errors. Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab.Universally used for all types of microbiological control testing.