QuickSwitch™ Quant H-2 Kb Tetramer Kit-BV421, MBL International
Supplier: MBL International
TB-7300-K4
103500-058EA 2307.69 USD
103500-058
QuickSwitch™ Quant H-2 Kb Tetramer Kit-BV421, MBL International
Assays Immunological Assays
QuickSwitch™ tetramer Kit utilizes a patented technique for exchanging up to ten peptides on an MHC class I tetramer. Reduce time and cost with this kit to create your own custom tetramers. The resulting custom tetramers can be used for multiple applications, including screening immunogenic peptides.
  • Kit Components:
  • Tetramer with an irrelevant exchangeable peptide in the MHC groove
  • A peptide exchange factor for catalyzing the peptide exchange reaction
  • A high affinity MHC-binding reference peptide used as peptide exchange positive control
  • FITC conjugated antibody specific of the exiting peptide
  • Magnetic beads conjugated with anti MHC antibody for tetramer capture
  • An Assay Buffer for diluting reagents and washing steps
Research Area: Immunology, Immune Monitoring
Quantification Included