Bovine/Calf Blood In Heparin Sodium, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
R500-0050 R500-0100
10800-434EA 228.81 USD
10800-434 10800-436
Bovine/Calf Blood In Heparin Sodium, Rockland Immunochemicals
Blood
Bovine/Calf Blood In Heparin Sodium
  • aseptically collected and prepared to order
  • blood draw in Anticoagulant
Bovine/calf whole blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Bovine/calf red blood cells are perishable and are collected and processed upon receipt of your order.
Washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.