Sterile Human Blood In Sodium Citrate, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
R214-0050
75932-278EA 652.99 USD
75932-278
Sterile Human Blood In Sodium Citrate, Rockland Immunochemicals
Blood
Sterile Human Blood In Sodium Citrate
  • aseptically collected and prepared to order
  • blood draw in Anticoagulant
Citrated human blood is aseptically collected and prepared to order from our colony of animals. Citrated human blood is ideal for blood agar plate preparations. Blood Agar is a general purpose enriched medium often used to grow fastidious organisms and to differentiate bacteria based on their hemolytic properties. Citrated human RBCs are also useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Human red blood cells are perishable and are collected and processed upon receipt of your order.
Blood agar plates, washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.