Human TIMP-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0312
76236-786EA 636 USD
76236-786
Human TIMP-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human TIMP-1 AccuSignal ELISA Kit
  • Natural and recombinant human TIMP-1.
  • There is no detectable cross-reactivity with other relevant proteins
  • .Expression system for standard: NSO; Immunogen sequence: C24-A207
The tissue inhibitor of metalloproteinases 1(TIMP1) is also called erythroid-potentiating activity(EPA). The X-linked gene for human TIMP1 is expressed in some but not all inactive X-containing somatic-cell hybrids, suggesting that this gene is either prone to reactivation or variable in its inactivation. Purified EPA specifically stimulates human and murine cells of the erythroid lineage, unlike murine interleukin-3(IL-3) which stimulates precursor cells from all haematopoietic lineages. TIMP1 is thought to play a regulatory role in connective tissues by forming inactive complexes with those metalloproteinases that are normally responsible for connective tissue turnover. The human gene encoding TIMP has been mapped to the X chromosome in the region Xp11.1-p11.4. The standard product used in this kit is natural TIMP-1 with the molecular mass of 22KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human TIMP-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA) or saliva to each empty well. It is recommended that each human TIMP-1 standard solution and each sample be measured in duplicate.