Human IL-1 beta ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0209
76236-642EA 636 USD
76236-642
Human IL-1 beta ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-1 beta AccuSignal ELISA Kit
  • Natural and recombinant human IL-1 beta.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A117-S269
Interleukin-1beta(IL-1beta) is a potent stimulator of bone resorption whose gene is mapped to 2q14, and has been implicated in the pathogenesis of high bone turnover and osteoporosis. IL-1beta, a prominent microglia-derived cytokine, caused oligodendrocyte death in coculture with astrocytes and microglia, but not in pure culture of oligodendrocytes alone. It also can cause nuclear export of a specific NCOR corepressor complex, resulting in derepression of a specific subset of nuclear factor-kappa-B(NFKB)-regulated genes. Furthermore, Microenvironmental IL-1beta and, to a lesser extent, IL-1alpha are required for in vivo angiogenesis and invasiveness of different tumor cells. Additional, the cooperation of IL-1beta and PDGFB induces contractile-to-synthetic phenotype modulation of human aortic smooth muscle cells in culture. Moreover, the association with disease may be explained by the biologic properties of IL-1beta, which is an important proinflammatory cytokine and a powerful inhibitor of gastric acid secretion.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 16.5pg/ml, 7.8pg/ml, 3.9pg/ml human IL-1 betastandard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernatants, serum or plasma(heparin, EDTA, citrate) to each empty well. We recommend that each human IL-1 betastandard solution and each sample is measured in duplicate.