Guinea Pig Gamma Globulin Fraction
Supplier: Rockland Immunochemicals
D605-0100
75928-296EA 314.48 USD
75928-296
Guinea Pig Gamma Globulin Fraction
Antibodies
Lyophilized Gamma Globulin Fraction
  • prepared from normal serum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography
  • control reagent
  • Western Blotting and ELISA experiments
Guinea Pig Gamma Globulin Fraction consists of the fraction of blood serum which contains whole antibodies as well as other non-albumin plasma proteins. Gamma globulins can be utilized therapeutically to temporarily boost an patient's immunity (such as after an immunosuppressive infection) or to increase the likelihood of kidney transplant acceptance. Gamma Globulin Fraction can be utilized in molecular biology experiments as a control reagent.
Guinea Pig Gamma Globulin Fraction can be utilized in Western Blotting and ELISA experiments.