Monkey (Cynomolgus) Serum Sterile, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D116-00-2
75929-996EA 939.25 USD
75929-996
Monkey (Cynomolgus) Serum Sterile, Rockland Immunochemicals
Sera (Serum)
Sterile Serum
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • separated from cellular components and purified through centrifugation and filtration techniques
Monkey Serum is used as a supplement to cell culture media. Monkey Serum is also suitable for use as a component of bioassays, immunoassays or enzyme assays. Monkey Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation. Monkey Serum is ideal for investigators in Cancer, Immunology, and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal