Cutters, Supreme, JH Williams Tool Group
Supplier: Williams
7292 7291 7190
76240-064EA 142.31 USD
76240-064 76238-880 76242-440
Cutters, Supreme, JH Williams Tool Group
Cutters
The precision screw joint helps to minimize the friction and maximize the alignment of cutting edges.
  • Polished and precision induction hardened to 58–60 HRC
  • Cutting capacity is calculated for solid copper wire
Features with ESD safe handles in synthetic material with leaf springs.