Tube Brushes, Short Handle Black Bristle, Torrington Brush Works
Supplier: TORRINGTON BRUSH WORKS INC. TE
06130 06131 06128 06129 06127
76239-534EA 7.57 USD
76239-534 76239-536 76240-264 76240-266 76240-594
Tube Brushes, Short Handle Black Bristle, Torrington Brush Works
Brushes
Black bristle tube or hole cleaning brushes. Bristle is resistant to most liquids except acids.
Uses for cleaning grime or loose debris from tubes or through holes.