Platinum-Cured Silicone Tubing, Newage Industries
Supplier: NewAge Industries
APSH-DB-0500L
76240-196EA 6575.93 USD
76240-196
Platinum-Cured Silicone Tubing, Newage Industries
Tubing Transfer Tubing
APSH-DB Polyester double braided silicone hose alternative to mandrel-built wrapped silicone hose, it provides several advantages over wrapped hose. It offers more flexibility and comparable working pressure ratings.
  • Extremely flexible with excellent bend radii
  • Less costly than wrapped-style silicone hose
  • Sterilizable/autoclavable
  • Imparts no taste or odor to critical streams
  • Manufactured and packaged in a Class 7 (Class 10,000) ISO-certified clean room
  • Documented lot traceable with identification on bags
Low volatile grade suitable for pharmaceutical and biomedical applications. Core made of silicone certified by the National Sanitation Foundation for food equipment materials (NSF-51). It resists temperature extremes, compression set, chemical attack, ozone, radiation, moisture, and environmental exposure.
Temperature range: –100 to +400 °F (–73.3 °C to +204.4 °C).
Certifications: USP Class VI, FDA CFR 177.2600, ISO 10993, European Pharmacopoeia 3.1.9, NSF 51, and 3A standards.
Caution: It is not intended for implantation and is not to be used for continuous steam applications.