Soldering Iron Handle, Heavy Duty, JBC
Supplier: JBC
T470-A
76241-434EA 395.95 USD
76241-434
Soldering Iron Handle, Heavy Duty, JBC
Soldering Supplies Soldering Irons
This soldering iron handle is perfect for intensive soldering jobs requiring continued high thermal power such as large ground planes, production of solar panels, armor and other applications.
  • Larger width grip and a screw which fixes the cartridge to prevent its rotation
It can provide up to 250 W when used with a HDE station and C470 cartridges.