Lindstrom 80 Cutters, JH Williams Tool Group
Supplier: Williams
8160 8148 8144 8142 8146 8151 8164 8150 8147 8211 8152
76238-892EA 429.26 USD
76238-892 76240-450 76238-894 76238-904 76238-890 76238-900 76238-902 76238-896 76238-898 76240-062 76238-888
Lindstrom 80 Cutters, JH Williams Tool Group
Cutters
Lindstrom 80 Series Cutters are made from high performance alloy steel.
  • Exacting induction hardened edges 63 - 65 HRC
  • Cutting capacity is listed for solid copper wire
These cutters are features with ESD safe handles in synthetic material with leaf springs. The inclusion of precision screw joint helps in minimizes friction and maximizes alignment of cutting edges.