10–in–1 Environmental Meter
Supplier: NEW GENIE GROUP INC SE
EN510
470310-234EA 270.15 USD
470310-234
10–in–1 Environmental Meter
Weather Stations
Convenient 10-in-1 Environmental Meter measuring Air Velocity, Air Flow, Air Temperature, Type K Temperature, Heat Index, Humidity, Wet Bulb, Dew Point, Windchill, and Light Level.
  • Measures Air Velocity, Air Flow, Temperature (Air/Type K), Humidity, Wet Bulb, Dew Point, Heat Index, Wind Chill and Light
  • Replaceable low friction ball bearing mini vane wheel to retain high accuracy
  • Measure light levels in Foot-Candle (Fc) or Lux units using a precision photo diode with cosine and color correction filter
  • Backlit display to view in dimly lit areas
  • Data Hold and Min/Max functions
  • Auto power off with disable
  • Complete with wrist strap, general purpose Type K bead wire probe, and three AAA batteries
The Type K input accepts standard Type K Thermocouple probes to measure surface temperature up to 2372 °F (1300 °C). Features large dual LCD with backlighting, Data Hold, and Min/Max function.
Delivery information: Supplied with built-in low friction ball bearing mini vane wheel, a precision photo diode with cosine and color correction filter, and a capacitive humidity sensor for high accuracy.