Vernier Go Direct® Dynamics Cart and Track System
Supplier: Vernier Software & Technology
DTS-GDX DTS-GDX-LONG
470310-204EA 635.99 USD
470310-204 470310-206
Vernier Go Direct® Dynamics Cart and Track System
Electrodes Other Electrodes
This completely wireless system simplifies experiment setup and allows basic experiments to be conducted with or without the track.
  • Position resolution: 0.25 mm, reported by default at 1 mm steps
  • Force range ±50 N
  • Accelerometer range ±160 m/s²
The Dynamics cart and track system with Go Direct Sensor cart includes essential laboratory equipment for teaching dynamics and kinematics. Using our Go Direct Sensor cart, students can explore force, position, velocity, and acceleration directly on their Bluetooth®-enabled device - no wires or additional equipment required. Each cart features built-in sensors to simplify experiment setup and allow experiments to be conducted on or off the track.
Delivery information: Supplied with Go Direct Sensor carts (1 green, 1 yellow), combination track/optics bench, adjustable end stop, adjustable two foot leveler, ultra pulley, pulley bracket, rod clamp, go direct sensor cart accessory kit, heavy hoop spring bumper (1), light hoop spring bumper (1), magnet tabs (4), hook-and-pile tabs (2), masses (4), magnetic bumper (2), force sensor hook (3), rubber bumper (3), anti-roll pegs (3).