Saturix Sterile Isopropyl Alcohol Spray Bottle, USP Grade
Supplier: FG Cleanwipes
Saturix
6LS7030VSIPA32B
76247-550CS 848.86 USD
76247-550
Saturix Sterile Isopropyl Alcohol Spray Bottle, USP Grade
Cleaning Agents Disinfectants
Packaged in no-leak, non-atomizing spray bottles, this blend of 70% IPA and 30% USP quality water is so pure and low in chemical extractibles that it can be used to remove residues from other cleaning solutions and solvents.
  • 30% USP purified DI water/70% USP grade Isopropyl alcohol blend filtered to 0.2 µm
  • Each bottle is double bagged
  • Gamma irradiated and validated to a sterility level of 10⁻⁶
  • Certificate of sterility provided with each case
  • Leak-tight spray bottle construction
  • Non-atomizing spray bottle nozzles
  • Pyrogen-free
  • Lot traceable
Applications include surface cleaning, use in USP <797> aseptic areas, use on gloved hands in sterile areas, use in critical medical device, biotechnology and pharmaceutical manufacturing areas, or wipe downs for pass-through controlled areas. Ideally suited for use with FG Clean Wipes sterile dry cleanroom wipes. This spray is also well suited for the decontamination of surfaces such as countertops, pass-throughs, and hoods. Recommended for use in ISO 3 - 8 controlled environments.
Spray bottles provide custom engineered performance and safety, and are supplied fully assembled and ready to use. This formulation eliminates mixing and processing in-house, while providing quick evaporation and leaving extremely low residues.
Certifications: Certificate of Sterility Included.