ChemMax® 2 Chemical Protective Disposable Coverall with Respirator-Fit Hood, Storm Flap, Elastic Wrists and Attached Boots, Lakeland Industries
Supplier: Lakeland
C2T151-2X C2T151-4X
76299-544CS 408.4 USD
76299-544 76299-546 76299-548 76299-550 76299-552 76299-554 76299-556
ChemMax® 2 Chemical Protective Disposable Coverall with Respirator-Fit Hood, Storm Flap, Elastic Wrists and Attached Boots, Lakeland Industries
Disposable Garments
Chemical Protective Disposable Coveralls are made with Saranex® 23P barrier film and heat sealed seams for a high degeree of chemical protection.
  • Saranex® 23P chemical barrier
  • Respirator fit hood
  • Heat sealed seams
Lakeland ChemMax® 2 garments are made with Saranex® 23P barrier film to provide the proven protection of an advanced chemical protective barrier. These garments provide a high level of chemical protection.
ChemMax® 2 is a superior and economical chemical protective offering from Lakeland, and is useful in protecting against hazardous chemicals and contaminents in the workplace.