Kanamycin sulfate, white to off-white powder USP
Supplier: TEKNOVA INC
|
|
Synonyms:
Kanamycin Monosulphate
, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
, Kanamycin A
, Kanamycin Monosulfate
, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulfate
K2105
K2105
100218-998EA
53.88
USD
100218-998
CA100218-998
Kanamycin sulfate, white to off-white powder USP
Kanamycin sulphate
Formula:
C₁₈H₃₈N₄O₁₅S MW: 582.6 g/mol |
MDL Number:
MFCD00070253 CAS Number: 25389-94-0 |
Specification Test Results
Potency |
|
pH (1%, H₂O) | 6.5 - 8.5 |
Moisture |
|
Ash |
|
Loss on Drying | 4.0% |
Residue on Ignition | 1.0% |
Chromatographic Purity | Passes |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...