PureFlo® Gas Disc Filter Capsules, Zenpure Americas
Supplier: ZENPURE AMERICAS C/O ST GOBAIN TE
G50CF0202H2H-1 G50CGH2H2H G50CF0202H2H-1
76240-988CS 1347.32 USD
76240-988 76248-234 76240-989
PureFlo® Gas Disc Filter Capsules, Zenpure Americas
Filters Capsule Filters
PureFlo® G50C disc capsules have been specially designed for simple, quick and efficient filtration of gases. All-polypropylene or polyethylene shell construction for excellent chemical, gas compatibility.
  • Hydrophobic barrier to protect from viral and bacterial contamination
  • Provides an ideal effective barrier for vacuum/vent protection from bio-hazardous contamination
The small, compact design of the filter capsule allows for superior air flow rate and low backpressure. Ideal for vacuum pump protection from contamination. A hydrophobic 1 µm or smaller micron size should be used for the application.
Certifications: Meets the 29CFR1910.1030 OSHA blood borne pathogen standard by serving as an 'Engineering Control' to reduce human exposure.