Ethylene glycol ≥99%, Reagent Grade
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Monoethylene glycol, Ethane-1,2-diol, 2-Hydroxyethanol, Ethylene alcohol, Ethanediol, Glycol alcohol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, 1,2-Dihydroxyethane, Glycol, Dihydroxyethane
102466-1L
102466-20L
102466-30KG
102466-5L
102466-4X2.5L
BJ102466-1LEA
141.83
USD
BJ102466-1L
BJ102466-2.5L
BJ102466-20L
BJ102466-225KG
BJ102466-30KG
BJ102466-5L
BJ102466-4X2.5L
Ethylene glycol ≥99%, Reagent Grade
Ethylene glycol
Formula:
HOCH₂CH₂OH Boiling Pt: 197 °C Melting Pt: –12 °C Density: 1.11 g/cm³ (20 °C) Flash Pt: 111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002885 CAS Number: 107-21-1 EINECS: 203-473-3 |
|
Specification Test Results
Refractive Index | n20/D 1.431 (lit.) |
Assay (GC) | Min. 99.0 % |
IR spectrum | complying |
APHA | Max. 10 |
Vapor Density | 2.1 (vs air) |
Explosion Limit | 15.3 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...