Anti-IL6R Rabbit Monoclonal Antibody [clone:033] APC (Allophycocyanin)

Supplier: Sino Biological
50280-R033-A-25 50280-R033-A-100
103629-990EA 90.9 USD
103629-990 103632-166
Anti-IL6R Rabbit Monoclonal Antibody [clone:033] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-6R / CD126 (rM IL-6R / CD126; Catalog#50280-M08H; NP_034689.2; Ile44-Glu357) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: IL-6R
Clonality: Monoclonal
Clone: Clone: 033
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR