Anti-SELL Rabbit Monoclonal Antibody [clone:414] APC (Allophycocyanin)

Supplier: Sino Biological
50045-R414-A-25 50045-R414-A-100
103629-904EA 70.7 USD
103629-904 103631-916
Anti-SELL Rabbit Monoclonal Antibody [clone:414] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SELL / CD62L / L-selectin (rM SELL / CD62L / L-selectin; Catalog#50045-M08H; NP_035476.1; Met 1-Asn 332) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: Clone: 414
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR