Anti-CD93 Mouse Monoclonal Antibody [clone:01] APC (Allophycocyanin)

Supplier: Sino Biological
12589-MM01-A-25 12589-MM01-A-100
103631-404EA 90.9 USD
103631-404 103632-020
Anti-CD93 Mouse Monoclonal Antibody [clone:01] APC (Allophycocyanin)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD93 / C1qR (rh CD93 / C1qR; Catalog#12589-H08H; Q9NPY3; Met1-Lys580) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: CD93
Clonality: Monoclonal
Clone: Clone: 01
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR