Anti-M13 Mouse Monoclonal Antibody [clone:MM05]

Supplier: Sino Biological
11973-MM05T-100 11973-MM05T-20
103629-688EA 270.69 USD
103629-688 103629-690 103636-118 76777-614
Anti-M13 Mouse Monoclonal Antibody [clone:MM05]
Antibodies
This product is a recombinant monoclonal antibody expressed from HEK293 cells.

Type: Primary
Antigen: M13
Clonality: Monoclonal
Clone: MM05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR