Anti-SLAMF6 Rabbit Monoclonal Antibody [clone:105] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
11945-R105-F-25 11945-R105-F-100
103629-678EA 70.7 USD
103629-678 103631-860
Anti-SLAMF6 Rabbit Monoclonal Antibody [clone:105] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human Ly108/SLAMF6 (rh Ly108/SLAMF6; Catalog#11945-H08H; Q96DU3-1; Met1-Met226) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: SLAMF6
Clonality: Monoclonal
Clone: Clone: 105
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR