Anti-CCL7 Mouse Monoclonal Antibody [clone:18] PE (Phycoerythrin)

Supplier: Sino Biological
11926-MM18-P-25 11926-MM18-P-100
103629-666EA 111.1 USD
103629-666 103631-848
Anti-CCL7 Mouse Monoclonal Antibody [clone:18] PE (Phycoerythrin)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MCP-3 / CCL7 (rh MCP-3 / CCL7; Catalog#11926-H08E; NP_006264.2; Gln24-Leu99) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: MCP3
Clonality: Monoclonal
Clone: Clone: 18
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR