Anti-OLFM4 Mouse Monoclonal Antibody [clone: 12]

Supplier: Sino Biological
11639-MM12-100 11639-MM12-20
103634-732EA 270.59 USD
103634-732 103634-734 103645-374 76777-458
Anti-OLFM4 Mouse Monoclonal Antibody [clone: 12]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human OLFM4 / GW112 (rh OLFM4 / GW112; Catalog#11639-H08H; NP_006409.3; Met 1-Gln 510).

Type: Primary
Antigen: OLFM4
Clonality: Monoclonal
Clone: 12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR