Anti-MST1R Mouse Monoclonal Antibody [clone:03] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
11608-MM03-F-25 11608-MM03-F-100
103629-586EA 70.7 USD
103629-586 103631-756
Anti-MST1R Mouse Monoclonal Antibody [clone:03] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD136/MST1R (rh CD136/MST1R; Catalog#11608-H08H; Q04912; Met1-Leu571) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: RON
Clonality: Monoclonal
Clone: Clone: 03
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR