Anti-ICOSLG Mouse Monoclonal Antibody [clone:01] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
11559-MM01-F-25 11559-MM01-F-100
103629-572EA 70.7 USD
103629-572 103631-742
Anti-ICOSLG Mouse Monoclonal Antibody [clone:01] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ICOS Ligand / B7-H2 (rh ICOS Ligand / B7-H2; Catalog#11559-H08H; NP_056074.1; Met1-Ser258) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: ICOS ligand
Clonality: Monoclonal
Clone: Clone: 01
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR