Anti-IL7R Rabbit Monoclonal Antibody [clone: 001] (PE (Phycoerythrin))

Supplier: Sino Biological
10975-R001-P-25 10975-R001-P-100
103629-306EA 90.9 USD
103629-306 103631-026
Anti-IL7R Rabbit Monoclonal Antibody [clone: 001] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL7R / IL-7R / CD127 (rh IL7R / IL-7R / CD127; Catalog#10975-H08H; P16871-1; Met1-Gly236) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: IL17RA
Clonality: Monoclonal
Clone: Clone: 001
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR