Anti-CD48 Rabbit Monoclonal Antibody [clone: 248] (APC (Allophycocyanin))

Supplier: Sino Biological
10797-R248-A-25 10797-R248-A-100
103629-208EA 90.9 USD
103629-208 103630-946
Anti-CD48 Rabbit Monoclonal Antibody [clone: 248] (APC (Allophycocyanin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD48 / SLAMF2 (rh CD48 / SLAMF2; Catalog#10797-H08H; NP_001769.2; Met 1-Ser 220) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: Anti-CD48 Antibody
Clonality: Monoclonal
Clone: Clone: 248
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR