Anti-CD14 Mouse Monoclonal Antibody [clone: 06] (PE (Phycoerythrin))

Supplier: Sino Biological
10073-MM06-P-25 10073-MM06-P-100
103628-750EA 70.7 USD
103628-750 103630-420
Anti-CD14 Mouse Monoclonal Antibody [clone: 06] (PE (Phycoerythrin))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD14 (rh CD14; Catalog#10073-H08H; NP_000582.1; Met1-Met344) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: Clone: 06
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR