Anti-SPARCL1 Rabbit Monoclonal Antibody [clone: 390] (PE (Phycoerythrin))

Supplier: Sino Biological
10046-R390-P-25 10046-R390-P-100
103628-716EA 111.1 USD
103628-716 103630-402
Anti-SPARCL1 Rabbit Monoclonal Antibody [clone: 390] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human SPARCL1 (rh SPARCL1; Catalog#10046-H08H; NP_004675.3; Met 1-Phe 664) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: SPARCL1
Clonality: Monoclonal
Clone: Clone: 390
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR