Anti-SPARCL1 Mouse Monoclonal Antibody [clone: 06] (PE (Phycoerythrin))

Supplier: Sino Biological
10046-MM06-P-25 10046-MM06-P-100
103628-712EA 111.1 USD
103628-712 103630-398
Anti-SPARCL1 Mouse Monoclonal Antibody [clone: 06] (PE (Phycoerythrin))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SPARCL1 (rh SPARCL1; Catalog#10046-H08H; NP_004675.3; Met 1-Phe 664) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: SPARCL1
Clonality: Monoclonal
Clone: Clone: 06
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR