Anti-SPARCL1 Mouse Monoclonal Antibody [clone: 05] (FITC (Fluorescein Isothiocyanate))

Supplier: Sino Biological
10046-MM05-F-25 10046-MM05-F-100
103628-708EA 90.9 USD
103628-708 103630-394
Anti-SPARCL1 Mouse Monoclonal Antibody [clone: 05] (FITC (Fluorescein Isothiocyanate))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SPARCL1 (rh SPARCL1; Catalog#10046-H08H; NP_004675.3; Met1-Phe664) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: SPARCL1
Clonality: Monoclonal
Clone: Clone: 05
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR