Anti-ERBB2 Rabbit Monoclonal Antibody [clone: 511] (PE (Phycoerythrin))

Supplier: Sino Biological
10004-R511-P-25 10004-R511-P-100
103628-464EA 90.9 USD
103628-464 103630-330
Anti-ERBB2 Rabbit Monoclonal Antibody [clone: 511] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, human cell-derived, recombinant human ErbB2 / HER2 extracellular domain (rh ErbB2; Catalog#10004-H08H; NP_004439.2; Met 1-Thr 652) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: Her2 / ERBB2
Clonality: Monoclonal
Clone: Clone: 511
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR