Nexel® Black Epoxy Wire Shelving, Global Industrial
Supplier: GLOBAL INDUSTRIAL TE
B970189 S2442B
76249-674EA 590.55 USD
76249-674 76253-212
Nexel® Black Epoxy Wire Shelving, Global Industrial
Furniture Shelves
The unique baked-on process bonds a black epoxy powder coat finish directly to the steel's surface resulting in an attractive, rugged and affordable shelving unit.
  • Open shelving permits sprinkler, air and light penetration and minimizes dust accumulation
  • High performance finish guaranteed against corrosion in dry storage applications
  • Easy assembly
These epoxy wire shelves feature a patented truss reinforced design for extra strength and greater capacities. Shelves easily adjust without tools and align accurately at 1" increments on numbered posts.
Certifications: NSF approved for dry use.
Delivery information: Starter unit includes four posts with leveling feet, four wire shelves and plastic snap on sleeves to secure shelves to posts. Individual shelves include snap-on sleeves.