Conductive Assembly Trays, Jensen Tools
Supplier: Jensen Tools + Supply
333000-5167 318000
76253-304EA 79.68 USD
76253-304 76253-102
Conductive Assembly Trays, Jensen Tools
Trays/Dishes/Pots Laboratory Trays/Dishes
Trays are made of reinforced thermoset polyester composite material (FibreStat 2000). Effective ESD protection for transporting components and assemblies.
  • Surface resistivity of <105 to 106 ohms
  • Resistance to abrasion, chemicals and solder
  • Ideal to carry heavy loads
Clean in steam or washer with a temperature range of –40 to 250 °F.