AlphaTec® Solvex® 37-155 Unsupported Nitrile Gloves, Ansell
Supplier: Ansell Healthcare
117145 117142
76238-046CS 709.02 USD
76238-046 76239-938
AlphaTec® Solvex® 37-155 Unsupported Nitrile Gloves, Ansell
Gloves
These gloves are made of a high-performance nitrile compound which provides an outstanding combination of strength and chemical resistance.
  • Great strength and chemical resistance
  • Ideal for working with petroleum solvents, caustics and animal fats
  • Embossed grip
When the gloves exposed to aromatic and petroleum solvents, as well as caustics and animal fats, it have proven to be a superior choice over rubber or neoprene. The gloves won’t swell, weaken, or degrade and they offer better protection against snags, punctures, abrasions, and cuts. And, as added benefits and it do not promote contact dermatitis, and their component materials.