AdvantaFlex® BioPharmaceutical Grade Tubing, NewAge Industries
Supplier: NewAge Industries
APAF-BP-0188-0313L APAF-BP-0125-0250L
76243-664EA 165.85 USD
76243-664 76240-788
AdvantaFlex® BioPharmaceutical Grade Tubing, NewAge Industries
Tubing Transfer Tubing
TPE (thermoplastic elastomer) tubing addresses the need for a flexible, translucent, sterilizable, moldable, heat sealable, and weldable biopharmaceutical tubing for fluid processing. AdvantaFlex maintains its physical properties following sterilization processes, resists kinking, remains translucent for visible product flow, and does not become gummy.
  • Sterile weldable and heat sealable
  • Available as leak-proof molded connections and tubing
  • Made from FDA-approved ingredients
  • Certified free of silicone oils and animal-derived ingredients
  • Sterilizable by autoclave or gamma radiation
  • Excellent low absorption and adsorption, low permeability
Available with validated sterility assurance of 10 to 6 via gamma irradiation per ISO 11137 method VDmax 25.
Smooth interior for excellent flow and performance. Translucent for fluid flow visibility. Excellent tubing component for Single-Use systems.
Single resin validation no need to graft to other tubing materials like silicone for media filling and sampling that involves peristaltic pumps; helps eliminate operator errors.
Used for sterile filling, vaccine production, pharmaceutical sampling and delivery systems, single-use systems, bioreactor processes, cell media, harvesting and fermentation, pharmaceutical production and processing, high purity water transfer filtration.
Certifications: Meets various ISO and USP standards, including Class VI. Meets European Pharmacopoeia 3.2.2.1 standards. REACH, RoHS and California Proposition 65 compliant. Meets ISO 9001:2015.
Packaging: Double bagged; sealed in polybags at manufacture for cleanliness. Documented lot traceable with identification on packaging.