Indicator Labels, Shockwatch™, Uline
Supplier: Uline
S-5159
76245-064BX 606.62 USD
76245-064
Indicator Labels, Shockwatch™, Uline
Labels
These labels indicates possible damage during shipping.
  • Non-resettable
  • Vial turns red to show impact
  • Activates automatically when dropped
Large size alerts handlers and is a permanent record of rough handling.