Rechargeable Battery Pack, Brady
Supplier: Brady Worldwide
114885
76247-306EA 332.53 USD
76247-306
Rechargeable Battery Pack, Brady
Batteries
Rechargeable battery pack, optimize the operation of your BMP®71 label printer with this spare or replacement NiMH battery.
  • Long battery life - prints up to 3900 1×2" labels per charge
The printer can be run while battery is charging. Enables cordless printer operation so you can print labels where you need them, when you need them.
Use AC adapter/charger unit (not included) to charge battery.
Delivery information: Package includes spare Printer Battery only. (label printer and charger sold separately)